missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E500, 99.0-100.5%, powder, Honeywell Fluka™
BIOPHARMA

Product Code. 15681310 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
6 x 1 kg
Unit Size:
1kg
5kg
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15681310

Brand: Honeywell-Fluka 134336X1KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E500, 99.0-100.5%, powder

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Quantity 6 x 1 kg
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Formula Weight 84.01
Percent Purity 99.0 to 100.5%
Grade puriss.
Chemical Name or Material Sodium Bicarbonate, Meets analytical specification of Ph. Eur.,BP, USP, FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.