missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(2-methoxyethoxy)aluminum hydride, 70 wt% solution in toluene (ca. 3.5M), AcroSeal™

Product Code. 10185847 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10185847 100 mL AcroSeal™ Glass bottle 100mL
10074627 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10185847 Supplier Thermo Scientific Supplier No. 430911000
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 22722-98-1 ∣ C6H16AlNaO4 ∣ 202.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22722-98-1 , 108-88-3
Molecular Formula C6H16AlNaO4
Molecular Weight (g/mol) 202.16
MDL Number MFCD00011631
Synonym Sodium dihydro-bis-(2-methoxyethoxy)aluminate in toluene

Specifications

Boiling Point 110.0°C
Color Amber to Colorless
Physical Form Viscous Liquid
Quantity 100 mL
Flash Point 4°C
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.036
Density 1.0360g/mL
Linear Formula [(CH3OCH2CH2O)2AlH2]Na
Merck Index 15,8702
Solubility Information Solubility in water: reacts. Other solubilities: soluble in aromatic hydrocarbons and ethers
Formula Weight 202.16
Percent Purity 68.5 to 71.5%
Grade Ungraded
Chemical Name or Material Sodium bis(2-methoxyethoxy)Aluminum hydride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium bis(2-methoxyethoxy)aluminiumhydride, 70 wt% sol. in toluene (ca. 3.5M)
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.