missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(trifluoromethylsulfonyl)imide

Product Code. 15482727
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15482727 250 mg 250mg
15462727 1 g 1g
15472727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15482727 Fornitore Thermo Scientific Alfa Aesar N. del fornitore 046640.MD
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 91742-21-1 | C2HF6NNaO4S2 | 304.133 g/mol

Sodium bis(trifluoromethylsulfonyl)imide is used as an electrolyte in batteries and fuel cells. It also finds use as an ionic liquid and Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 91742-21-1
Molecular Formula C2HF6NNaO4S2
Molecular Weight (g/mol) 304.133
MDL Number MFCD14156021
InChI Key QXZNUMVOKMLCEX-UHFFFAOYSA-N
Synonym Sodium triflimide; Sodium trifluoromethanesulfonimide
PubChem CID 131887538
IUPAC Name sodium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Na]

Specifica

Melting Point 257°C to 258°C
Linear Formula (CF3SO2)2NNa
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 303.13
Physical Form Solid
Chemical Name or Material Sodium bis (trifluoromethylsulfonyl)imide

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.