missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(trimethylsilyl)amide, 1M soln. in THF

Product Code. 11342706
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342706 100 mL 100mL
11352706 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342706 Supplier Thermo Scientific Alfa Aesar Supplier No. L08779.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1070-89-9 | C6H18NNaSi2 | 183.377 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bis(trimethylsilyl)amide is useful for the generation of carbenes, the amination of sulfonamides, and for the preparation of lanthanide complexes. It is significantly accelerated the polymerization of pheneylacetylene in conjunction with rhodium (I) catalysis. It is used to deprotonate ketones and esters to generate enolate derivatives, phosphonium salts, generating Wittig reagents.

Solubility
Soluble in benzene, toluene and ether. Reacts with water.

Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents, Oxygen. Store away from water and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-89-9
Molecular Formula C6H18NNaSi2
Molecular Weight (g/mol) 183.377
MDL Number MFCD00009835
InChI Key WRIKHQLVHPKCJU-UHFFFAOYSA-N
Synonym sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
PubChem CID 2724254
IUPAC Name sodium;bis(trimethylsilyl)azanide
SMILES C[Si](C)(C)[N-][Si](C)(C)C.[Na+]

Specifications

Quantity 100 mL
UN Number UN2924
Beilstein 3629917
Flash Point −17°C (1°F)
Solubility Information Soluble in benzene,toluene and ether. Reacts with water.
Formula Weight 183.38
Concentration or Composition (by Analyte or Components) 1M soln. in THF
Sensitivity Moisture sensitive; May form a precipitate
Density 0.904
Chemical Name or Material Sodium bis(trimethylsilyl)amide
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium bis(trimethylsilyl)amide, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Tetrahydrofuran, Sodium bis(trimethylsilyl)amide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.