missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(trimethylsilyl)amide, 95+%, pure

Product Code. 10246510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246510 5 g Glass bottle 5g
10144890 25 g Glass bottle 25g
10712384 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10246510 Supplier Thermo Scientific Acros Supplier No. 215960050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1070-89-9 | C6H18NNaSi2 | 183.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-89-9
Molecular Formula C6H18NNaSi2
Molecular Weight (g/mol) 183.38
MDL Number MFCD00009835
InChI Key WRIKHQLVHPKCJU-UHFFFAOYSA-N
Synonym sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
PubChem CID 2724254
IUPAC Name sodium;bis(trimethylsilyl)azanide
SMILES C[Si](C)(C)[N-][Si](C)(C)C.[Na+]

Specifications

Melting Point 171°C to 175°C
Color Yellow to Beige
Density .916g/mL
Infrared Spectrum Authentic
Assay Percent Range 95+%
Packaging Glass bottle
Linear Formula [(CH3)3Si]2NNa
Quantity 5 g
Fieser 01,1046; 03,261; 04,442; 06,529; 12,441; 16,307
Solubility Information Solubility in water: reacts
Formula Weight 183.38
Percent Purity 95+%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Sodium bis(trimethylsilyl)amide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium bis(trimethylsilyl)amide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.