missing translation for 'onlineSavingsMsg'
Learn More

Sodium bismuth oxide, 80%

Product Code. 11448006
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448006 500 g 500g
11428006 25 g 25g
11438006 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448006 Supplier Thermo Scientific Alfa Aesar Supplier No. A14073.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12232-99-4 | BiNaO3 | 279.967 g/mol

Sodium bismuthate oxide is used as an oxidant in the preparation of hexavalent Americium (Am) during the separation of Am from the lanthanides., hydrothermal synthesis of bismuth oxychloride (BiOCl) micrometer-sized single crystals. It is also used as a precursor in the synthesis of bismuth nanoforms under microwave (MW) irradiation. It used to converts almost any manganese compound to permanganate, which is easily assayed spectrophotometrically.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bismuthate oxide is used as an oxidant in the preparation of hexavalent Americium (Am) during the separation of Am from the lanthanides., hydrothermal synthesis of bismuth oxychloride (BiOCl) micrometer-sized single crystals. It is also used as a precursor in the synthesis of bismuth nanoforms under microwave (MW) irradiation. It used to converts almost any manganese compound to permanganate, which is easily assayed spectrophotometrically.

Solubility
Soluble in aqueous acid. Insoluble in water.

Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12232-99-4
Molecular Formula BiNaO3
Molecular Weight (g/mol) 279.967
MDL Number MFCD00003474
InChI Key PNYYBUOBTVHFDN-UHFFFAOYSA-N
Synonym sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium
PubChem CID 4063671
IUPAC Name sodium;oxido(dioxo)bismuth
SMILES [O-][Bi](=O)=O.[Na+]

Specifications

Quantity 500 g
Merck Index 14,8585
Solubility Information Soluble in aqueous acid. Insoluble in water.
Formula Weight 279.97
Percent Purity 80%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Sodium bismuth oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.