missing translation for 'onlineSavingsMsg'
Learn More

Sodium bismuth oxide, ACS, 80% min

Product Code. 11377528
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377528 25 g 25g
11387528 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377528 Supplier Thermo Scientific Alfa Aesar Supplier No. 033219.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manganese testing in iron, steel, ores | CAS: 12232-99-4 | BiNaO3 | 279.967 g/mol

Sodium bismuth oxide is used for the determination of manganese in iron, steel and ores. It is also used in the separation of plutonium and acts as a strong oxidizer. It is used in the conversion of manganese compound to permanganate. Further, it is used in the preparation of hexavalent americium (Am) during its separation from the lanthanides and in the hydrothermal synthesis of bismuth oxychloride micrometer-sized single crystals. In addition to this, it is employed as a precursor in the preparation of bismuth nanoforms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12232-99-4
Molecular Formula BiNaO3
Molecular Weight (g/mol) 279.967
MDL Number MFCD00003474
InChI Key PNYYBUOBTVHFDN-UHFFFAOYSA-N
Synonym sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium
PubChem CID 4063671
IUPAC Name sodium;oxido(dioxo)bismuth
SMILES [O-][Bi](=O)=O.[Na+]

Specifications

Color Yellow to Brown
Physical Form Powder
Quantity 25 g
Merck Index 14,8585
Solubility Information Insoluble in cold water; Decomposes in hot water forming Bi2O3,NaOH,and oxygenInsoluble in water.
Formula Weight 279.97
Percent Purity ≤80%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Sodium bismuth oxide
Show More Show Less
compliance-icons
Product Identifier
  • Sodium bismuth oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.