missing translation for 'onlineSavingsMsg'
Learn More

Sodium bismuthate(V), 85%

Product Code. 11904341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11904341 1 kg Glass bottle 1kg
10012770 25 g Glass bottle 25g
10195020 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11904341 Supplier Thermo Scientific Acros Supplier No. 205210010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12232-99-4 | BiNaO3 | 279.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12232-99-4
Molecular Formula BiNaO3
Molecular Weight (g/mol) 279.96
InChI Key PNYYBUOBTVHFDN-UHFFFAOYSA-N
Synonym sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium
PubChem CID 4063671
IUPAC Name sodium;oxido(dioxo)bismuth
SMILES [O-][Bi](=O)=O.[Na+]

Specifications

Color Yellow to Brown
Physical Form Powder
Quantity 1 kg
Assay Percent Range 80% min. (Iodometry)
Linear Formula NaBiO3
Merck Index 15,8721
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes in hot water and acids
Formula Weight 279.96
Percent Purity 85%
Packaging Glass bottle
Chemical Name or Material Sodium bismuthate(V)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.