missing translation for 'onlineSavingsMsg'
Learn More

Sodium bisulfite solution, Purum, ∼40%, Honeywell Fluka™

Product Code. p-7071808 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Unit Size:
1 L
2.5 L
25 L
Product Code. Quantity unitSize
15691310 1 L 1 L
15601320 2.5 L 2.5 L
15611320 25 L 25 L
3 options
This item is not returnable. View return policy

Product Code. 15691310

Brand: Honeywell-Fluka 134381L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum, ∼40%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-90-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
MDL Number MFCD00003530
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
Synonym sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
PubChem CID 23665763
ChEBI CHEBI:26709
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

Quantity 1 L
Linear Formula NaHSO3
UN Number 3082
Formula Weight 104.06g/mol
Grade purum
Concentration or Composition (by Analyte or Components) 40%
Chemical Name or Material Sodium Bisulfite Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.