missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate ACS AR Granular, Macron Fine Chemicals™

Product Code. 15548754
Change view
Click to view available options
Quantity:
2.5 kg
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15548754 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548754 Supplier Avantor Macron Supplier No. 745706
  • ONLINE EXCLUSIVE PRICE
    390.86 €
    353.28 € / 2.5kg
    Save 37.58 €
    9% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1303-96-4
Physical Form Granules
Quantity 2.5 kg
Molecular Formula Na2B4O7·10H2O
Formula Weight 381.37
Grade ACS
Packaging Poly
Chemical Name or Material Sodium Borate, 10-Hydrate
compliance-icons
Product Identifier
  • di-Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: di-Sodium tetraborate decahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.