missing translation for 'onlineSavingsMsg'
Learn More

Sodium borodeuteride, 98 atom% D, for NMR

Product Code. 10338430 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10338430 1 g Glass Bottle 1g
10256630 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10338430 Supplier Thermo Scientific Acros Supplier No. 194950010
  • ONLINE EXCLUSIVE PRICE
    75.10 €
    72.90 € / 1g
    Save 2.20 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 15681-89-7 | BH4Na | 37.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15681-89-7
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003519
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym sodium borodeuteride, sodium tetradeuterioboron, sodium borohydride-d4, sodium 2h4 tetrahydroborate 1-, sulfatase, borate 1-, tetrahydro-d4-, sodium, nabd4, sodium 2h4 boranuide, deuterated sodium borohydride
PubChem CID 23673181
SMILES [BH4-].[Na+]

Specifications

Physical Form Powder and/or Chunks
Color Gray to White
Deuterium (D) 97.5% min.
Packaging Glass Bottle
Quantity 1 g
Melting Point >300.0°C
Assay Percent Range 98 atom% D
Linear Formula NaBD4
Fieser 03,281
IUPAC Name sodium boranuide
Formula Weight 41.86
Percent Purity 98 atom% D
Grade NMR
Chemical Name or Material Sodium borodeuteride, 98 atom % D
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium borodeuteride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.