missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 12% solution in 40% aq. NaOH solution, AcroSeal™

Product Code. 10119689 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10119689 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10119689 Supplier Thermo Scientific Acros Supplier No. 429131000
  • ONLINE EXCLUSIVE PRICE
    94.50 €
    88.20 € / 100mL
    Save 6.30 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16940-66-2 ∣ BH4Na ∣ 37.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 130.0°C to 135.0°C
Chemical Name or Material Sodium borohydride
CAS 16940-66-2, 1310-73-2, 67-56-1, 7732-18-5
Color Colorless
Physical Form Liquid
Quantity 100 mL
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.4
Density 1.4000g/mL
Molecular Formula BH4Na
Linear Formula NaBH4
MDL Number MFCD00003518
Merck Index 15,8727
Synonym Sodium tetrahydroborate, SBH
Solubility Information Solubility in water: completely soluble
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES [BH4-].[Na+]
IUPAC Name sodium boranuide
Molecular Weight (g/mol) 37.83
Formula Weight 37.82
Percent Purity 11 to 13%
Grade Ungraded
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.