missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 98%, granules, ca 3 mm

Product Code. 10422895 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10422895 500 g Plastic Bottle 500g
10617591 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10422895 Supplier Thermo Scientific Acros Supplier No. 419475000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16940-66-2 | BH4Na | 37.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym Sodium tetrahydroborate, SBH
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Melting Point 360.0°C
Color White
Physical Form Granules
Quantity 500 g
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula NaBH4
UN Number 1426
Fieser 01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189
Merck Index 15, 8727
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ammonia- reacts with acids,soluble in amines,pyridine and dimethylether of,diethylene glycol,insol.in other ethers,hydrocarb.,alkyl chlorides,reacts with oxidizing agents
Formula Weight 37.83
Percent Purity 98%
Packaging Plastic Bottle
Chemical Name or Material Sodium borohydride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium borohydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H301-Toxic if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.