missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 98% min

Product Code. 11369359
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11369359 500 g 500g
11359359 100 g 100g
2 missing translation for 'options'
Questo articolo non è restituibile. Consulta la politica di reso

Codice prodotto. 11369359

missing translation for 'mfr': Thermo Scientific Alfa Aesar 088983.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso

CAS: 16940-66-2 | BH4Na | 37.83 g/mol

Sodium borohydride is used as a reagent in the reduction of amino acids and their derivatives. Also used as a catalyst in the ammonia borane dehydrogenation. Sodium borohydride is used as a reducing agent in the synthesis of gold nanoparticles. It is also used for wood pulp and as a blowing agent for plastics. It reduces the aldehyde and keto groups to give the corresponding alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
Synonym Sodium tetrahydridoborate
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifica

Melting Point 400°C (decomposition)
Color White
Physical Form Granules
Quantity 500 g
Assay Percent Range 98% min.
Solubility Information Soluble in water. Also soluble in liquid ammonia,amines,pyridine.
Formula Weight 37.83
Ignition Point 220°C (428°F)
Odor Odorless
Sensitivity Moisture sensitive
Density 1.074 g/cm3 at 20°C
Chemical Name or Material Sodium borohydride
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.