Learn More
Sodium borohydride, 99%, (VenPureTM AF caplets), caplets
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 360.0°C |
| Physical Form | Caplets |
| Quantity | 500 g |
| Packaging | Plastic Bottle |
| Assay Percent Range | 99% |
| Linear Formula | NaBH4 |
| Merck Index | 15,8727 |
| Solubility Information | Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents |
| Formula Weight | 37.83 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
- Sodium borohydride
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 C
- Substance which, in contact with water, emits flammable gases Category 1
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- H301-Toxic if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- EUH014-Reacts violently with water.
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.