missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borohydride-d4, TRC

Product Code. 30416573
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416573 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30416573 Supplier Toronto Research Chemicals Supplier No. TRCS61490225G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Borohydride-d4
CAS 15681-89-7
Molecular Formula Na . B 2H4
Quantity 25 g
Synonym Sodium Borodeuteride, Sodium Tetrahydro-d4-borate, Sodium Tetradeuterioborate, Sodium Tetradeuteroborate, Sodium Tetrahydroborate-d4
InChI Formula InChI=1S/BH4.Na/h1H4;/q-1;+1/i1D4;
SMILES [Na+].[2H][B-]([2H])([2H])[2H]
IUPAC Name sodium;tetradeuterioboranuide
Molecular Weight (g/mol) 41.86
Formula Weight 42.0555
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.