Learn More
Sodium borohydride, stable aq. soln., 4.4M in 14M NaOH
Descrizione
Sodium borohydride is used as a reagent in the reduction of amino acids and their derivatives. Also used as a catalyst in ammonia borane dehydrogenation. It acts as a reducing agent in the synthesis of gold nanoparticles. It is also used for wood pulp and as a blowing agent for plastics. It reduces aldehyde and keto groups to give the corresponding alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 2 L |
| Sensitivity | Air Sensitive |
| Density | 1.4 g/mL |
| Linear Formula | NaBH4 |
| UN Number | UN3320 |
| Solubility Information | Miscible with water,liquid ammonia,amines and pyridine. |
| Formula Weight | 37.83 |
| Grade | Ungraded |
| Vedi altri risultati |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.