missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromate, 99.5% (metals basis)

Product Code. 11357697
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
This item is not returnable. View return policy

Product Code. 11357697

Brand: Thermo Scientific Alfa Aesar 014101.A3

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-38-0 | BrNaO3 | 150.891 g/mol

Sodium bromate is used as an oxidant to convert tetrahydropyranyl ethers to carbonyl compounds. It acts as an analytical reagent. It is also used for dissolving gold from its ores, boiler cleaning, oxidation of sulfur and vat dyes. Further, it is used in hair treatment formulations as a hair-permanent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-38-0
Molecular Formula BrNaO3
Molecular Weight (g/mol) 150.891
MDL Number MFCD00003476
InChI Key XUXNAKZDHHEHPC-UHFFFAOYSA-M
Synonym sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
PubChem CID 23668195
ChEBI CHEBI:75229
IUPAC Name sodium;bromate
SMILES [O-]Br(=O)=O.[Na+]

Specifications

Melting Point 381°C (decomposition)
Color White
Physical Form Crystals
Quantity 2 kg
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water (2.5 parts in cold water,1.1 part in boiling water)Soluble in water and ammonia. Insoluble in ethanol,acetone and ether.
Formula Weight 150.9
Odor Odorless
Refractive Index 1.594
Appearance White crystalline
Density 3.34
Chemical Name or Material Sodium bromate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Ihr Feedback wurde übermittelt