missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromate, 99+%, extra pure

Product Code. 10072730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10072730 100 g Glass bottle 100g
10082730 500 g Glass bottle 500g
13488458 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10072730 Supplier Thermo Scientific Acros Supplier No. 208871000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-38-0 | BrNaO3 | 150.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-38-0
Molecular Formula BrNaO3
Molecular Weight (g/mol) 150.9
MDL Number MFCD00003476
InChI Key XUXNAKZDHHEHPC-UHFFFAOYSA-M
Synonym sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
PubChem CID 23668195
ChEBI CHEBI:75229
IUPAC Name sodium;bromate
SMILES [O-]Br(=O)=O.[Na+]

Specifications

Melting Point 381.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 100 g
Assay Percent Range 99+%
Linear Formula NaBrO3
Fieser 01,1055
Merck Index 15,8728
Solubility Information Solubility in water: 364g/L (20°C). Other solubilities: insoluble in alcohol , ether and aceton
Formula Weight 150.9
Percent Purity ≥99%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 3.33
Density 3.3300g/mL
Chemical Name or Material Sodium bromate, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium bromate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.