missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bromide, Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
500g
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 2.5 kg |
| Linear Formula | NaBr |
| UN Number | NONH for all modes of transport |
| IUPAC Name | sodium;bromide |
| Formula Weight | 102.89g/mol |
| Percent Purity | 99 to 100.5% (calc. to the dried substance) |
| Grade | puriss. |
| Packaging | Plastic Bottle |
| Vapor Pressure | 1mmHg (806°C) |
| Chemical Name or Material | Sodium Bromide, Meets analytical specification of Ph. Eur.BP,USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction