missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Sodium butyrate

Product Code. 18003922
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18003922 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18003922 Supplier Tocris Bioscience™ Supplier No. 3850/50
  • ONLINE EXCLUSIVE PRICE
    108.00 €
    101.85 € / 50mg
    Save 6.15 €
    5% Off
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Histone deacetylase inhibitor

Sodium butyrate is a histone deacetylase inhibitor (HDAC). Sodium butyrate restores contextual memory in a transgenic mouse model of Alzheimer's disease. In a transgenic mouse model of Huntington's disease, sodium butyrate ameliorates the neurodegenerative phenotype and extends survival. Sodium butyrate induces pancreatic progenitor formation, and directs the differentiation of mouse embryonic stem cells into hepatocytes when used in combination with the cytokine Activin A. Sodium butyrate also augments transcription factor stimulated iPSC generation. In hepatocellular carcinoma cells, Sodium butyrate downregulates expression of c-Myc resulting in decreased hexokinase 2 levels, inhibition of aerobic glycolysis and reduced cell growth.
TRUSTED_SUSTAINABILITY

Specifications

CAS 156-54-7
Quantity 50 mg
Molecular Formula C4H7NaO2
Formula Weight 110.09
Purity >98%
Molecular Weight (g/mol) 110.09
Chemical Name or Material Sodium butyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.