missing translation for 'onlineSavingsMsg'
Learn More

Sodium cacodylate, 0.1M buffer soln., pH 6.5

Product Code. 15453149
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453149 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15453149 Supplier Thermo Scientific Alfa Aesar Supplier No. J60367.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 6131-99-3214.024 g/mol

Sodium cacodylate, is used as an arsenate compound for proteomics research. It is used as a buffer in electron microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium cacodylate, is used as an arsenate compound for proteomics research. It is used as a buffer in electron microscopy.

Solubility
Soluble in water (∼100 g/L) at 20°C, ethanol (400 mg/mL), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-99-3
Molecular Weight (g/mol) 214.024

Specifications

Concentration 0.1M buffer soln.
pH 6.5
Physical Form Liquid
Quantity 100 mL
compliance-icons
Product Identifier
  • Sodium cacodylate, 0.1M buffer solution, pH 6.5
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Arsenate, dimethyl,sodium, Hydrochloricacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.