missing translation for 'onlineSavingsMsg'
Learn More

Sodium caprylate, 98%

Product Code. 10418302 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10418302 100 g 100g
10419262 500 g 500g
10617164 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10418302 Supplier Thermo Scientific Acros Supplier No. 334601000
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1984-06-1 | C8H15NaO2 | 166.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1984-06-1
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.2
MDL Number MFCD00058511
InChI Key BYKRNSHANADUFY-UHFFFAOYSA-M
Synonym sodium octanoate, sodium caprylate, octanoic acid, sodium salt, sodium n-octanoate, sodium octoate, natrium octanoat, caprylic acid sodium salt, octanoic acid sodium salt, octanoic acid, sodium salt 1:1, unii-9xtm81vk2b
PubChem CID 23664772

Specifications

Melting Point >225°C
Color White
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 100 g
Solubility Information Solubility in water: soluble in water
Formula Weight 166.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sodium caprylate, 98%
compliance-icons
Product Identifier
  • Sodium caprylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.