missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, 0.05N Standardized Solution

Product Code. 15461347
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461347 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15461347 Supplier Thermo Scientific Alfa Aesar Supplier No. 035609.K2
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 497-19-8 ∣ CNa2O3 ∣ 105.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 497-19-8
Physical Form Liquid
Quantity 1 L
Molecular Formula CNa2O3
MDL Number MFCD00003494
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C([O-])=O
IUPAC Name disodium carbonate
Molecular Weight (g/mol) 105.99
PubChem CID 10340
ChEBI CHEBI:29377
Formula Weight 105.99
Grade Ungraded
Appearance Colourless liquid
Chemical Name or Material Sodium carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.