missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, 99.999%, (trace metal basis)

Product Code. 11954301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11954301 5 g Glass bottle 5g
11964301 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11954301 Supplier Thermo Scientific Acros Supplier No. 436800050
  • ONLINE EXCLUSIVE PRICE
    48.45 €
    47.00 € / 5g
    Save 1.45 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Boiling Point 1600.0°C
Melting Point 851.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99.999%
Linear Formula Na2CO3
Merck Index 15,8731
Solubility Information Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
IUPAC Name disodium carbonate
Formula Weight 105.99
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Sodium carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.