missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Anhydrous, Certified AR for Analysis, Fisher Chemical™

Product Code. 10498030 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
2.5kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10498030 5 kg Plastic keg 5kg
10264540 500 g Plastic powder jar 500g
10548070 1 kg Plastic powder jar 1kg
10518260 2.5 kg Plastic powder jar 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10498030 Supplier Fisher Chemical Supplier No. S/2920/65
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium Carbonate Anhydrous, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Boiling Point 1600°C
Melting Point 854°C
Color White
pH 11.3
Physical Form Solid
Quantity 5 kg
IUPAC Name disodium carbonate
Formula Weight 105.99g/mol
Packaging Plastic keg
compliance-icons
Product Identifier
  • Sodium carbonate anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.