missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Decahydrate, ≥99.0% (T), Honeywell Fluka™

Product Code. 15666760 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
5kg
Product Code. Quantity Packaging unitSize
15666760 250 g Poly bottle 250g
15656760 1 kg Poly bottle 1kg
15676760 5 kg Poly bottle 5kg
3 options
This item is not returnable. View return policy

Product Code. 15666760

Brand: Honeywell-Fluka 71360250G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a.,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-1-6132
Molecular Formula CH20Na2O13
Molecular Weight (g/mol) 286.14
MDL Number MFCD00149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 32°C
Color Colorless
pH 11.6 (at 20°C)
Physical Form Solid
Quantity 250 g
Linear Formula Na2CO3 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 286.14g/mol
Percent Purity ≥99.0% (T)
Odor Odorless
Packaging Poly bottle
Density 1.460g/cm3
Chemical Name or Material Sodium carbonate decahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.