missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Decahydrate, ≥99.0% (T), Honeywell Fluka™

Product Code. 15676760 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
5kg
Product Code. Quantity Packaging unitSize
15676760 5 kg Poly bottle 5kg
15666760 250 g Poly bottle 250g
15656760 1 kg Poly bottle 1kg
3 options
This item is not returnable. View return policy

Product Code. 15676760

Brand: Honeywell-Fluka 713605KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a.,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 2-1-6132
Molecular Formula CH20Na2O13
Molecular Weight (g/mol) 286.14
MDL Number MFCD00149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifica

Melting Point 32°C
Color Colorless
pH 11.6 (at 20°C)
Physical Form Solid
Quantity 5 kg
Linear Formula Na2CO3 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 286.14g/mol
Percent Purity ≥99.0% (T)
Odor Odorless
Packaging Poly bottle
Density 1.460g/cm3
Chemical Name or Material Sodium carbonate decahydrate
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.