missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate decahydrate, 99+%, for analysis

Product Code. 10400301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10400301 2.5 kg Plastic bottle 2.5kg
10348110 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10400301 Supplier Thermo Scientific Acros Supplier No. 213500025
  • ONLINE EXCLUSIVE PRICE
    96.25 €
    90.20 € / 2.5kg
    Save 6.05 €
    6% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6132-02-1 | CNa2O3·10H2O | 286.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-1-6132
Molecular Formula CNa2O3·10H2O
Molecular Weight (g/mol) 286.14
MDL Number MFCD00149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 34°C
Color White
Physical Form Small Crystals
Quantity 2.5 kg
Packaging Plastic bottle
Specific Gravity 1.46
Assay Percent Range 99+%
Density 1.4600g/mL
Linear Formula Na2CO3·10H2O
Fieser 07,322
Merck Index 15, 8731
Solubility Information Solubility in water: 210g/L (20°C). Other solubilities: soluble in glycerol,insoluble in alcohol
Formula Weight 286.14
Percent Purity ≥99%
Grade Analytical
Chemical Name or Material Sodium carbonate decahydrate, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Sodium carbonate decahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.