missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate decahydrate, 99+%, for biochemistry, Thermo Scientific Chemicals

Product Code. 12641256 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
Product Code. Quantity unitSize
12641256 2.5 kg 2.5kg
12604937 500 g 500g
2 options
This item is not returnable. View return policy

Product Code. 12641256

Brand: Thermo Scientific Acros 125020025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-1-6132
Molecular Formula CNa2O3·10H2O
Molecular Weight (g/mol) 286.14
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 34.0°C
Quantity 2.5 kg
Assay Percent Range 99+%
Linear Formula Na2CO3·10H2O
Merck Index 15,8731
Solubility Information Clear colorless
Formula Weight 286.14
Percent Purity ≥99%
Grade Biochemical
Specific Gravity 1.46
Density 1.4600g/mL
Chemical Name or Material Sodium carbonate decahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.