missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate decahydrate, 99+%, extra pure

Product Code. 11944301 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
10 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
100g
10kg
This item is not returnable. View return policy

Product Code. 11944301

Brand: Thermo Scientific Acros 271070100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2-1-6132 | CNa2O3·10H2O | 286.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-1-6132
Molecular Formula CNa2O3·10H2O
Molecular Weight (g/mol) 286.14
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 34.0°C
Color White
Physical Form Crystals
Quantity 10 kg
Assay Percent Range 99+%
Linear Formula Na2CO3·10H2O
Fieser 7322
Merck Index 158731
Solubility Information Solubility in water: 210g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic Drum
Specific Gravity 1.46
Density 1.46 g/mL
Chemical Name or Material Sodium carbonate decahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.