missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate decahydrate, 99+%, extra pure

Product Code. 10636144 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
100g
10kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10636144 100 g Plastic Bottle 100g
10033350 2.5 kg Plastic Bottle 2.5kg
11944301 10 kg Plastic Drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10636144 Supplier Thermo Scientific Acros Supplier No. 271071000
  • ONLINE EXCLUSIVE PRICE
    22.80 €
    21.40 € / 100g
    Save 1.40 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2-1-6132 | CNa2O3·10H2O | 286.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-1-6132
Molecular Formula CNa2O3·10H2O
Molecular Weight (g/mol) 286.14
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 34.0°C
Color White
Physical Form Crystals
Quantity 100 g
Packaging Plastic Bottle
Specific Gravity 1.46
Assay Percent Range 99+%
Density 1.46 g/mL
Linear Formula Na2CO3·10H2O
Fieser 7322
Merck Index 158731
Solubility Information Solubility in water: 210g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Formula Weight 286.14
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Sodium carbonate decahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.