missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate Decahydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10090640 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
2.5kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10090640 5 kg Plastic keg 5kg
10785531 1 kg Plastic powder jar 1kg
10264490 2.5 kg Plastic powder jar 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10090640 Supplier Fisher Chemical Supplier No. S/2720/65
  • ONLINE EXCLUSIVE PRICE
    593.00 €
    568.00 € / 5kg
    Save 25.00 €
    4% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2-1-6132
Molecular Formula CH20Na2O13
Molecular Weight (g/mol) 286.14
MDL Number 149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 34°C
Color White
pH 11 to 12
Physical Form Solid
Quantity 5 kg
Packaging Plastic keg
Formula Weight 286.01g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.