missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate decahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., E500, 99.5-100.5% (ex dried subst.), Solstice
BIOPHARMA

Product Code. 15671290
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
50 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
50kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15671290 1 kg 1kg
15681290 2.5 kg 2.5kg
15601300 6 x 1 kg Pack of 6
15691290 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15671290 Supplier Honeywell-Fluka Supplier No. 134141KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., E500, 99.5-100.5% (ex dried subst.)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-1-6132
Molecular Formula CH20Na2O13
Molecular Weight (g/mol) 286.14
MDL Number MFCD00149178
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

Quantity 1 kg
Linear Formula Na2CO3 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 286.14g/mol
Percent Purity 99.5 to 100.5% (ex dried subst.)
Grade puriss.
Chemical Name or Material Sodium Carbonate Decahydrate, Meets analytical specification of Ph. Eur.BP, FCC
compliance-icons
Product Identifier
  • Sodium carbonate decahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sodium carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.