missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate decahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., E500, 99.5-100.5% (ex dried subst.), Honeywell Fluka™
Shop All Honeywell Fluka Products

missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
2.5 kg
50 kg
6 x 1 kg
missing translation for 'unitSize'
1kg
2.5kg
50kg
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 2.5 kg |
| Linear Formula | Na2CO3 · 10H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 286.14g/mol |
| Percent Purity | 99.5 to 100.5% (ex dried subst.) |
| Grade | puriss. |
| Chemical Name or Material | Sodium Carbonate Decahydrate, Meets analytical specification of Ph. Eur.BP, FCC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction