missing translation for 'onlineSavingsMsg'
Learn More

Sodium carboxymethyl cellulose (Viscosity:800-1200 mPa.s), MedChemExpress

Product Code. 16412718
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16412718 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16412718 Supplier Medchem Express Supplier No. HYY0703

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Sodium carboxymethyl cellulose (Viscosity:800-1200 mPa.s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent, paste and barrier agent.

Sodium carboxymethyl cellulose (Viscosity:800-1200 mPa.s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent, paste and barrier agent.

Specifications

Chemical Name or Material Sodium carboxymethyl cellulose (Viscosity:800-1200 mPa.s)
CAS 9004-32-4
Color White
Purity Grade Notes Research
Quantity 5 g
Synonym CMC-Na (Viscosity:800-1200 mPa.s)
SMILES [Sodium carboxymethyl cellulose]
Grade Research
Physical Form Powder

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.