missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium caseinate, pract.

Product Code. 10772402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10772402 500 g Glass bottle 500g
10772212 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772402 Supplier Thermo Scientific Chemicals Supplier No. 419495000
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium caseinate, CAS # 9005-46-3, is the salt of the phosphoprotein class found naturally in milk and cheese. It contains all nine essential amino acids and may be used as a molecular reagent, or in cell culture research on growth and differentiation. | CAS: 9005-46-3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sodium caseinate is the salt of the phosphoprotein class known as caseins
  • Caseins are found in animal tissues, especially milk, and they contain all nine essential amino acids

Applications

  • Casein can be used in protease enzyme activity studies, in vitro experiments on cell growth and differentiation, or as a blocking solution for western blot
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-46-3
MDL Number MFCD00130736

Specifications

Color White to Cream
pH 6 to 7.5 (5% soln.)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500 g
Percent Purity ≥92% (protein) (on dry substance)
Physical Form Powder
Chemical Name or Material Sodium caseinate, pract.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.