missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride 0.9% (Normal Saline), USP, Sterile Grade, Cytiva Hyclone™

Product Code. 31937778
Change view
Click to view available options
Quantity:
6 x 500 mL
20 L
6 x 1 L
Packaging:
Bioprocess Bag
PET Plastic Bottle
Unit Size:
16 L
20 L
6 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31937778 6 x 500 mL PET Plastic Bottle 16 L
31937773 6 x 1 L PET Plastic Bottle 6 L
31937770 20 L Bioprocess Bag 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31937778 Supplier HyClone products Cytiva Supplier No. Z1376
  • ONLINE EXCLUSIVE PRICE
    132.00 €
    113.83 € / 16 L
    Save 18.17 €
    13% Off
 Restricted Item
Estimated Shipment: 25-02-2027
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium Chloride Injection, USP is a sterile solution of sodium chloride in water for injection. It contains no antimicrobial agents. It contains NLT 95.0% and NMT 105.0% of the labeled amount of sodium chloride (NaCl).

  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Sterile and endotoxin tested
  • Ideal for medical device and biopharmaceutical manufacturing
  • Suitable in research, testing, cell culture and molecular protocols
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Physical Form Liquid
Quantity 6 x 500 mL
Grade USP
Concentration or Composition (by Analyte or Components) 0.009
Packaging PET Plastic Bottle
Chemical Name or Material Sodium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.