missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, 5M aq. soln., RNAse free

Product Code. 15413209
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413209 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15413209 Supplier Thermo Scientific Alfa Aesar Supplier No. J60434.AK
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7647-14-5 ∣ ClNa ∣ 58.44 g/mol

Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media. a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications

Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media. a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.

Solubility

Soluble in water (358 mg/ml at 20°C), alcohol (very slightly), glycerol (100 mg/ml), and ammonia. Insoluble in hydrochloric acid.

Notes

Incompatible with strong oxidizing agents. Store in a cool, dry, well-ventilated area away from incompatible substances. Store protected from moisture.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7647-14-5
Physical Form Liquid
Quantity 250 mL
Molecular Formula ClNa
Linear Formula NaCl
MDL Number MFCD00003477
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
Solubility Information Soluble in water (358mg/ml at 20°C),alcohol (very slightly),glycerol (100mg/ml),and ammonia. Insoluble in hydrochloric acid.
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Formula Weight 58.44
Grade Ungraded
Concentration or Composition (by Analyte or Components) 5M aq. soln.
Chemical Name or Material Sodium chloride, RNAse free
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.