missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium chloride, 99.5%, for biochemistry

Product Code. 12685077 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12685077 5 kg Plastic bottle 5kg
12605087 250 g Plastic bottle 250g
12675077 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12685077 Supplier Thermo Scientific Chemicals Supplier No. 447300050
  • ONLINE EXCLUSIVE PRICE
    258.00 €
    242.00 € / 5kg
    Save 16.00 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7647-14-5 ∣ ClNa ∣ 58.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461.0°C
Melting Point 801.0°C
Physical Form Powder, Crystals and/or Chunks
Quantity 5 kg
Packaging Plastic bottle
Assay Percent Range 99.4% min. (Argentometry)
Linear Formula NaCl
Merck Index 15,8734
Solubility Information (0.1 M in water) Clear colorless
IUPAC Name sodium chloride
Formula Weight 58.44
Percent Purity 99.5%
Grade Biochemical
Chemical Name or Material Sodium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.