missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, meets analytical specification of Ph.Eur

Product Code. 11964909 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
3 kg
10 kg
25 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
10kg
1kg
25kg
3kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11964909 25 kg Plastic keg 25kg
11984051 1 kg Plastic powder jar 1kg
11994051 3 kg Plastic powder jar 3kg
11994929 10 kg Plastic keg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11964909 Supplier Fisher Chemical Supplier No. S/3161/70
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Quantity 25 kg
IUPAC Name sodium;chloride
Packaging Plastic keg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.