missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, ACS, ≥99%, Ultrapure

Product Code. 15855188 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
Packaging:
Plastic bottle
Unit Size:
10kg
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15855188 500 g Plastic bottle 500g
15805338 1 kg Plastic bottle 1kg
15815338 5 kg Plastic bottle 5kg
15825338 10 kg Plastic bottle 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15855188 Supplier Thermo Scientific Supplier No. J21618.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for precipitation of DNA | CAS: 7647-14-5 | ClNa | 58.44 g/mol

Used for the precipitation of DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1413°C
Melting Point 801°C
Physical Form Crystalline Powder
Quantity 500 g
Packaging Plastic bottle
Specific Gravity 2.165
Density 2.165
IUPAC Name sodium;chloride
Formula Weight 58.44
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Sodium Chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.