missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Cell Culture Reagent Grade, MP Biomedicals™

Product Code. 11482011 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
Unit Size:
10kg
1kg
500g
5kg

Product Code. 11482011

Brand: MP Biomedicals 02194738.5

Please to purchase this item. Need a web account? Register with us today!

Used in a wide variety of biochemical applications. Sodium Chloride, Cell Culture Reagent Grade, MP Biomedicals is a chemical found widely in nature. It is considered an essential nutrient. This product is used as a diluent, in protein crystallization, in

Sodium chloride is used in a wide variety of biochemical applications, including:

  • In protein crystallization
  • Intravenous fluids (normal saline is 0.9% w/v in water10)
  • Density gradients
  • A diluent to increase ionic strength in buffers or culture media
  • In salt-out procedures in the isolation of DNA
  • In high concentrations for preservation of foods, since bacteria cannot grow in high salt conditions

A salt-ice mixture in the ratio of approximately 33g sodium chloride to 100g ice (at −1°C) will drop in temperature to as low as &minus’21°C, depending on the rate of stirring and the size of the ice chunks. Excess amounts of sodium chloride can destroy electrolyte balance and cause death in most animals, including humans.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water (357mg/mL at 25°C; 384mg/mL at 100°C), glycerol (100mg/mL); very slightly soluble in ethanol. Almost insoluble in concentrated HCl.
IUPAC Name sodium chloride
Formula Weight 58.44g/mol
Percent Purity ≥99.5%
Grade Cell Culture Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.