missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride CertiFied AR, Fisher Chemical™

Product Code. 15526005 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15526005 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15526005 Supplier Fisher Chemical Supplier No. S/3160/53
  • ONLINE EXCLUSIVE PRICE
    30.10 €
    25.40 € / 500g
    Save 4.70 €
    15% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium chloride CertiFied AR 500g Meets EP specifications | CAS: 7647-14-5 | ClNa | 58.44 g/mol

Sodium chloride CertiFied AR 500g Meets EP specifications

  • Sodium chloride CertiFied AR 500g Meets EP specifications Plastic Bottle Cas No. 7647-14-5
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
Chemical Name or Material Sodium chloride
Melting Point 801°C
CAS 7647-14-5
Quantity 500 g
Packaging PLASTIC
Molecular Formula ClNa
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.