missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, crystalline powder PDV, 99+%

Product Code. p-7082058
Click to view available options
Quantity:
1000 g
25,000 g
5000 g
Unit Size:
1000g
25000g
5000g
This item is not returnable. View return policy

Product Code. 11455063

Brand: Thermo Scientific Alfa Aesar A12313.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-14-5 | ClNa | 58.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium chloride is widely used as a food additive, and a preservative. It is employed in the Chloralkali process for producing chlorine and sodium hydroxide, in the Solvay process for producing sodium carbonate and calcium chloride, in the Mannheim process and in the Hargreaves process for producing sodium sulfate and hydrochloric acid. It is used in the manufacture of sodium chlorate, water softening and de-icing of roadways. Further it is used in several industries like pharmaceutical, textile, dyeing, rubber food and agriculture. It is used in the processing of steel, copper, vanadium, and other metals.

Solubility
Freely soluble in water. Slightly soluble in alcohol and ammonia.

Notes
It is sensitive to moisture in air. It should be protected in an air tight container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1413°C
Melting Point 801°C
pH 6 to 9
Physical Form Crystalline Powder
Quantity 1000 g
Merck Index 14,8599
Solubility Information Freely soluble in water. Slightly soluble in alcohol and ammonia.
Formula Weight 58.44
Percent Purity ≥99%
Odor Odorless
Refractive Index 1.5442
Sensitivity Hygroscopic
Density 2.165
Chemical Name or Material Sodium chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.