missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Extra Pure, SLR, Fisher Chemical™

Product Code. 10616082 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Unit Size:
1kg
3kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10616082 500 g 500g
10598630 3 kg 3kg
10356340 5 kg 5kg
10112640 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10616082 Supplier Fisher Chemical Supplier No. S/3120/53
  • ONLINE EXCLUSIVE PRICE
    25.12 €
    18.60 € / 500g
    Save 6.52 €
    25% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 500 g
IUPAC Name sodium;chloride
Formula Weight 58.44g/mol
Packaging Plastic powder jar
Vapor Pressure 1mmHg 865
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.