missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, for HPLC, Certified, Fisher Chemical

Product Code. 10274392 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10274392 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Please note: Due to manufacturing changes this item is no longer EP compliant, for EP compliant Sodium Chloride please use the S/3161 counterpart
 
Product Code. 10274392 Supplier Fisher Chemical Supplier No. S/3165/53
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Please note: Due to manufacturing changes this item is no longer EP compliant, for EP compliant Sodium Chloride please use the S/3161 counterpart
 

Specifications

Boiling Point 1461°C
Melting Point 801°C
CAS 7647-14-5
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 500 g
Molecular Formula ClNa
MDL Number MFCD00003477
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium;chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Formula Weight 58.44g/mol
Packaging Plastic powder jar
Vapor Pressure 1mmHg 865
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.