missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride solution 1mol/L, Fisher Chemical™

Product Code. 12903614 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12903614 1000 mL Bottle 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12903614 Supplier Fisher Chemical Supplier No. VL 293 1000
  • ONLINE EXCLUSIVE PRICE
    140.00 €
    121.22 € / 1000mL
    Save 18.78 €
    13% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

CAS 7647-14-5, 7732-18-5
Quantity 1000 mL
Packaging Bottle
Molecular Formula ClNa
MDL Number MFCD00003477
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Concentration or Composition (by Analyte or Components) 1M (1N)
Chemical Name or Material Sodium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.