missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, USP, Fisher Chemical™

Product Code. 15989851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
3 kg
Unit Size:
3kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15989851 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15989851 Supplier Thermo Scientific Acros Supplier No. USS6403
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Boiling Point 1461°C (760 mmHg)
Melting Point 801°C
CAS 7647-14-5
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 3 kg
Molecular Formula ClNa
Linear Formula NaCl
MDL Number MFCD00003477
UN Number 5234
Merck Index 15, 8734
Solubility Information soluble in glycerol (1 g/10 mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Formula Weight 58.44
Percent Purity 99.0 to 100.5% (FCC)
Grade FCC/USP
Identification Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC)
Loss on Drying 0.5% max. (FCC), 0.5% max. (USP/FCC)
Packaging Plastic Bottle
Sensitivity Hygroscopic
Appearance white solid
Vapor Pressure 1 mmHg (at 440°C)
Density 2.165 g/cm3
Chemical Name or Material Sodium chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.