missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorodifluoroacetate, 97%

Product Code. 11442408
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442408 25 g 25g
11452408 100 g 100g
11462408 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442408 Supplier Thermo Scientific Alfa Aesar Supplier No. A15756.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1895-39-2 | C2ClF2NaO2 | 152.457 g/mol

It is used as pharmaceutical intermediate. Reactions of aldehydes with triphenylphosphine and sodium chlorodifluoroacetate yield 1,1-difluoro olefins. It is a general reaction and a one step synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Reactions of aldehydes with triphenylphosphine and sodium chlorodifluoroacetate yield 1,1-difluoro olefins. It is a general reaction and a one step synthesis.

Solubility
Soluble in dimethoxyethane, diglyme (0.60 g mL-1), triglyme, dimethyl formamide.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1895-39-2
Molecular Formula C2ClF2NaO2
Molecular Weight (g/mol) 152.457
MDL Number MFCD00064771
InChI Key MRTAVLDNYYEJHK-UHFFFAOYSA-M
Synonym sodium chlorodifluoroacetate, sodium 2-chloro-2,2-difluoroacetate, chlorodifluoroacetic acid sodium salt, sodium chloro difluoro acetate, acetic acid, chlorodifluoro-, sodium salt, sodiumchlorodifluoroacetate, difluorochloroacetic acid sodium salt, sodium difluorochloroacetate, chlorodifluoroacetic acid sodium
PubChem CID 2734985
IUPAC Name sodium;2-chloro-2,2-difluoroacetate
SMILES C(=O)(C(F)(F)Cl)[O-].[Na+]

Specifications

Melting Point ∼195°C (decomposition)
Quantity 25 g
Beilstein 4162363
Solubility Information Soluble in dimethoxyethane,diglyme (0.60 g mL-1),triglyme,dimethyl formamide.
Formula Weight 152.46
Percent Purity 97%
Chemical Name or Material Sodium chlorodifluoroacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.